Merck
  • Fabrication of a cytochrome c biosensor based on cytochrome oxidase/NiO-NPs/cMWCNT/PANI modified Au electrode.
Fabrication of a cytochrome c biosensor based on cytochrome oxidase/NiO-NPs/cMWCNT/PANI modified Au electrode.
Journal of biomedical nanotechnology (2013-04-30)
Bhawna Batra, Suman Lata, Sunita Rani, C S Pundir
ABSTRACT
An amperometric biosensor for determination of Cytochrome c (Cyt c) was fabricated by immobilizing Cytochrome c oxidase (COx) onto nickel oxide nanoparticles (NiO-NPs) decorated carboxylated multiwalled carbon nanotubes/polyaniline (NiO-NPs/cMWCNT/PANI) film electrodeposited on the surface of a gold (Au) electrode. The electrochemical characteristics of immobilized COx were investigated by cyclic voltammetry (CV), electrochemical impedance spectroscopy (EIS), Scanning Electron Microscopy (SEM) and Fourier transform Infra-red spectroscopy (FTIR). Cyclic voltammetric (CV) studies of the electrode at different stages of construction of enzyme electrode demonstrated that the modified Au electrode had enhanced electrochemical oxidation of H2O2, which offers a number of attractive features to develop amperometric biosensors based on split of H2O2. There was a good linear relationship between the current (mA) and Cyt c concentration in the range 5 x 10(-12) M to 5 x 10(-7) M. The sensor had a detection limit of 5 x 10(-12) M (S/N = 3) with a high sensitivity of 3.7 mA cm(-2) nM(-1). The sensor gave accurate and satisfactory results, when employed for determination of Cyt c in different serum samples.
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Nickel(II) oxide, green, −325 mesh, 99%
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Nickel(II) oxide, 99.99% trace metals basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Nickel(II) oxide, nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Nickel(II) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation