Merck
  • Use of microscope-integrated near-infrared indocyanine green videoangiography in the surgical treatment of intramedullary cavernous malformations: report of 8 cases.
Use of microscope-integrated near-infrared indocyanine green videoangiography in the surgical treatment of intramedullary cavernous malformations: report of 8 cases.
Journal of neurosurgery. Spine (2013-03-12)
Toshiki Endo, Misaki Aizawa-Kohama, Kenichi Nagamatsu, Kensuke Murakami, Akira Takahashi, Teiji Tominaga
ABSTRACT
The characteristics and efficacy of indocyanine green (ICG) videoangiography in cavernous malformation (CM) have not been fully elucidated. The purpose of this paper is to examine the potential utility of ICG videoangiography in the surgical treatment of intramedullary CMs. The authors conducted a retrospective review of 8 cases involving 5 men and 3 women who had undergone surgery for intramedullary CM between January 2008 and July 2011. All patients were evaluated by means of MRI. The MRI findings and clinical history in all cases suggested intramedullary CM as a preoperative diagnosis. In 2 of 8 cases, dilated venous structures associated with CMs were demonstrated. In one of these cases, there were coexisting extramedullary CMs. Intraoperatively, ICG fluorescence was observed for 5 minutes using microscope-integrated videoangiography. In all 8 cases, intra- and extramedullary CMs were seen as avascular areas on ICG videoangiography. Indocyanine green videoangiography helped surgeons to localize and predict margins of the lesions before performing myelotomy. Importantly, in the cases with associated venous anomalies, ICG videoangiography was useful in delineating and preserving the venous structures. In extramedullary CMs located dorsal to the spinal cord, gradual ICG infiltration was seen, starting at 110 seconds and maximal at 210 seconds after injection. Postoperative MRI confirmed total removal of the lesions in all cases, and subsequent recovery of all patients was uneventful. Indocyanine green videoangiography provided useful information with regard to the detection of lesion margins by demonstrating intramedullary CMs as avascular areas. In cases associated with venous anomalies, ICG contributed to safe and complete removal of the CMs by visualizing the venous structure. In extramedullary CMs, ICG videoangiography demonstrated the characteristic of slow blood flow within CMs.
Sigma-Aldrich
Cardiogreen, polymethine dye
Sigma-Aldrich
Cardiogreen, for microscopy