Skip to Content
End of year dates: Orders must be billed by 05/12. Merck Mexico will be closed from 18/12 to 02/01.
Merck
  • Orthogonal spin labeling and Gd(III)-nitroxide distance measurements on bacteriophage T4-lysozyme.
Orthogonal spin labeling and Gd(III)-nitroxide distance measurements on bacteriophage T4-lysozyme.
The journal of physical chemistry. B (2013-02-28)
Luca Garbuio, Enrica Bordignon, Evan K Brooks, Wayne L Hubbell, Gunnar Jeschke, Maxim Yulikov
ABSTRACT
We present the first example of chemoselective site-specific spin labeling of a monomeric protein with two spectroscopically orthogonal spin labels: a gadolinium(III) chelate complex and a nitroxide radical. A detailed analysis of the performance of two commercially available Gd(III) ligands in the Gd(III)-nitroxide pulse double electron-electron resonance (DEER or PELDOR) experiment is reported. A modification of the flip angle of the pump pulse in the Gd(III)-nitroxide DEER experiment is proposed to optimize sensitivity.
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
1 Estimated to ship on February 19, 2025
MXP 12,107.00