Merck
  • Towards a first-principles model of Fermi resonance in the alkyl CH stretch region: application to 1,2-diphenylethane and 2,2,2-paracyclophane.
Towards a first-principles model of Fermi resonance in the alkyl CH stretch region: application to 1,2-diphenylethane and 2,2,2-paracyclophane.
The Journal of chemical physics (2013-02-22)
Evan G Buchanan, Jacob C Dean, Timothy S Zwier, Edwin L Sibert
ABSTRACT
The spectroscopy of two flexible hydrocarbons, 1,2-diphenylethane (DPE) and 2,2,2-paracyclophane (TCP) is presented, and a predictive theoretical model for describing the alkyl CH stretch region of these hydrocarbons is developed. Ultraviolet hole-burning spectroscopy identified two isomers of DPE and a single conformation of TCP present in the supersonic jet expansion. Through the analysis of the ground state low-frequency vibronic spectroscopy obtained by dispersed fluorescence, conformational assignments were made for both DPE and TCP. The two isomers of DPE were found to retain the low energy structures of butane, being present in both the gauche and anti structures. TCP forms a C(2) symmetric structure, differing from the predicted lower energy C(3) conformation by the symmetry of the ethano bridges (-CH(2)CH(2)-) linking the phenyl substituents. Resonant ion-dip infrared spectroscopy is used to record single-conformation IR spectra of the two conformers of DPE and the single conformer of TCP in the alkyl CH stretch region and in the mid-IR that covers the CH bend fundamentals. A local mode Hamiltonian that incorporates cubic stretch-bend coupling is developed. Its parameters are obtained from density functional theory methods. Full dimensional calculations are compared to those that use reduced dimensional Hamiltonians in which anharmonic CH stretches and scissor modes are Fermi coupled. Excellent agreement is found. Scale factors of select terms in the reduced dimensional Hamiltonian are determined by fitting the theoretical Hamiltonian to the anti-DPE spectrum. The scaled Hamiltonian is then used to predict successfully structures for the remaining lower symmetry experimentally determined spectra in the alkyl CH stretch region.
Sigma-Aldrich
Bibenzyl, ReagentPlus®, 99%