Merck
  • Tourniquet-induced ischemia-reperfusion injuries during extremity surgery at children's age: impact of anesthetic chemical structure.
Tourniquet-induced ischemia-reperfusion injuries during extremity surgery at children's age: impact of anesthetic chemical structure.
Redox report : communications in free radical research (2013-02-12)
Ivana Budic, Dusica Pavlovic, Dusanka Kitic, Gordana Kocic, Tatjana Cvetkovic, Dusica Simic, Tatjana Jevtovic-Stoimenov
ABSTRACT
The aim of this study was to determine the relationship between the antioxidant profile of anesthetics and its relation to total antioxidant capacity (TAC) of plasma in children who underwent tourniquet-induced ischemia-reperfusion (IR) injury during extremity operations. Children were randomized into three groups: general inhalational anesthesia with sevoflurane (group S), total intravenous anesthesia (TIVA) with propofol (group T), and regional anesthesia (group R). Venous blood samples were obtained before peripheral nerve block and induction of general anesthesia (baseline), 1 minute before tourniquet release (BTR), and 5 and 20 minutes after tourniquet release (ATR). Plasma TAC as well as antioxidant potential of propofol, thiopental, and bupivacaine were measured using the 2,2-diphenyl-1-picrylhydrazyl (DPPH) assay. Plasma TAC in group T was increased significantly at 20 minutes ATR in comparison with basal and BTR values, and also was significantly higher in comparison with plasma TAC in groups S and R measured at the same time point. The radical scavenging activity of anesthetics in vitro indicated that only propofol possessed a significant antioxidative activity in the reaction with DPPH radical in comparison with thiopental and bupivacaine. These data confirm that TIVA with propofol attenuates oxidative stress related to tourniquet-induced ischaemia-reperfusion injury in children.
Product Description
Supelco
(±)Thiopental solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis