Merck
A new antenna system for microwave non-invasive hyperthermia lipolysis.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference (2013-02-01)
Sangbok Park, Joosung Hwang, Youngwoo Kwon, Changyul Cheon
ABSTRACT
In this paper, we present an antenna system for microwave non-invasive hyperthermia lipolysis. The antenna system consists of a circular waveguide antenna radiating electromagnetic waves, AlN (Aluminum Nitride) radome and heat sink. The AlN radome with heat sink helps to extract heat from the skin to keep skin temperature not to rise during heating the lipolysis. The antenna was designed to be operated with TE(21) mode to maintain uniform temperature over wider area. The usability of the proposed system was verified by performing numerical simulation and hyperthermia lipolysis experiments on rats.
Sigma-Aldrich
Aluminum nitride, nanopowder, <100 nm particle size
Sigma-Aldrich
Aluminum nitride, powder, 10 μm, ≥98%