Merck
  • Novel bisimidazolium pincers as low loading ligands for in situ palladium-catalyzed Suzuki-Miyaura reaction in the ambient atmosphere.
Novel bisimidazolium pincers as low loading ligands for in situ palladium-catalyzed Suzuki-Miyaura reaction in the ambient atmosphere.
Chemical communications (Cambridge, England) (2013-01-04)
Chao Gao, Hongjun Zhou, Siping Wei, Yinsong Zhao, Jingsong You, Ge Gao
ABSTRACT
A series of novel triazinonide-bridged bisimidazolium pincers were easily synthesized by quaternization of functionalized N-phenylimidazoles with highly reactive cyanuric chloride under mild conditions. The pincer was proven to be a very efficient ligand for in situ Pd-catalyzed Suzuki-Miyaura reaction with ppm-level catalyst loading.
Sigma-Aldrich
Cyanuric chloride, 99%