Merck
  • Application of carvedilol in a dog with pseudoephedrine toxicosis-induced tachycardia.
Application of carvedilol in a dog with pseudoephedrine toxicosis-induced tachycardia.
The Canadian veterinary journal = La revue veterinaire canadienne (2013-01-02)
Min-Hee Kang, Hee-Myung Park
ABSTRACT
A 15-year-old Yorkshire terrier dog was presented after ingesting 1 capsule of an over-the-counter cold medication containing pseudoephedrine (120 mg/capsule) and cetirizine (5 mg/capsule). Treatment was initiated with acepromazine and carvedilol. The dog responded well to treatment. This is the first known case report using carvedilol to control pseudoephedrine toxicosis.
Product Description
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acepromazine maleate, ≥98% (HPLC)
Sigma-Aldrich
Cetirizin dihydrochloride, ≥98.0% (HPLC)
Sigma-Aldrich
(1S,2S)-(+)-Pseudoephedrine, 98%