Merck
  • A protein from Pleurotus eryngii var. tuoliensis C.J. Mou with strong removal activity against the natural steroid hormone, estriol: purification, characterization, and identification as a laccase.
A protein from Pleurotus eryngii var. tuoliensis C.J. Mou with strong removal activity against the natural steroid hormone, estriol: purification, characterization, and identification as a laccase.
Enzyme and microbial technology (2012-10-09)
Mitsuhiro Ueda, Kayo Shintani, Akiko Nakanishi-Anjyuin, Masami Nakazawa, Mizuho Kusuda, Fumiki Nakatani, Takashi Kawaguchi, Sho-Ichi Tsujiyama, Masanobu Kawanishi, Takashi Yagi, Kazutaka Miyatake
ABSTRACT
A protein with strong removal activity against the natural estrogen estriol was purified from a culture supernatant of Pleurotus eryngii var. tuoliensis C.J. Mou. The protein was characterized as a laccase and had a molecular mass of 60kDa on SDS-PAGE. The enzyme was most active at pH 7.0 and 50°C. The partial N-terminal amino acid sequence of the enzyme showed homology with laccases from mushrooms, such as Pleurotus ostreatus, Coriolus versicolor (current name: Trametes versicolor), Pycnoporus cinnabarinus, and P. eryngii. A recombinant yeast assay confirmed that laccase treatment was very efficient for removing the estrogenic activity of steroid estrogens. Our results suggest that the enzyme may be applicable as a potential factor for removing natural steroid hormones.
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Estriol, meets USP testing specifications
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®