Merck
  • Biphenyl liquid crystalline epoxy resin as a low-shrinkage resin-based dental restorative nanocomposite.
Biphenyl liquid crystalline epoxy resin as a low-shrinkage resin-based dental restorative nanocomposite.
Acta biomaterialia (2012-07-31)
Sheng-Hao Hsu, Rung-Shu Chen, Yuan-Ling Chang, Min-Huey Chen, Kuo-Chung Cheng, Wei-Fang Su
ABSTRACT
Low-shrinkage resin-based photocurable liquid crystalline epoxy nanocomposite has been investigated with regard to its application as a dental restoration material. The nanocomposite consists of an organic matrix and an inorganic reinforcing filler. The organic matrix is made of liquid crystalline biphenyl epoxy resin (BP), an epoxy resin consisting of cyclohexylmethyl-3,4-epoxycyclohexanecarboxylate (ECH), the photoinitiator 4-octylphenyl phenyliodonium hexafluoroantimonate and the photosensitizer champhorquinone. The inorganic filler is silica nanoparticles (∼70-100 nm). The nanoparticles were modified by an epoxy silane of γ-glycidoxypropyltrimethoxysilane to be compatible with the organic matrix and to chemically bond with the organic matrix after photo curing. By incorporating the BP liquid crystalline (LC) epoxy resin into conventional ECH epoxy resin, the nanocomposite has improved hardness, flexural modulus, water absorption and coefficient of thermal expansion. Although the incorporation of silica filler may dilute the reinforcing effect of crystalline BP, a high silica filler content (∼42 vol.%) was found to increase the physical and chemical properties of the nanocomposite due to the formation of unique microstructures. The microstructure of nanoparticle embedded layers was observed in the nanocomposite using scanning and transmission electron microscopy. This unique microstructure indicates that the crystalline BP and nanoparticles support each other and result in outstanding mechanical properties. The crystalline BP in the LC epoxy resin-based nanocomposite was partially melted during exothermic photopolymerization, and the resin expanded via an order-to-disorder transition. Thus, the post-gelation shrinkage of the LC epoxy resin-based nanocomposite is greatly reduced, ∼50.6% less than in commercialized methacrylate resin-based composites. This LC epoxy nanocomposite demonstrates good physical and chemical properties and good biocompatibility, comparable to commercialized composites. The results indicate that this novel LC nanocomposite is worthy of development and has potential for further applications in clinical dentistry.
Sigma-Aldrich
Biphenyl, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%
Pricing and availability is not currently available.
Supelco
Biphenyl, PESTANAL®, analytical standard
Pricing and availability is not currently available.