Merck
  • [Genetic diversity and volatile components of Asarum sieboldii in Qin-ba region].
[Genetic diversity and volatile components of Asarum sieboldii in Qin-ba region].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-07-25)
Su-ying Hu, Gang He, Bei Chen, Lang-jun Cui, Jian-xiong Yang
ABSTRACT
To analyze the genetic diversity and the volatile components of Asarum sieboldii from seven habitats in Qin-ba region. The genetic diversity of the herb was analyzed by ISSR (inter simple sequence repeat) markers; The relative content volatile components of the herb were dectected by head space solid-phase microextraction gas chromatogrphy-mass spectrometry (HS-SPME-GC-MS). The contents of the 3 main components were analyzed by steam distillation gas chromatography-mass spectrometry (GC-MS). 57 bands were amplified from 7 populations by 6 reliable primers, 51 of which were polymorphic (89.47% of the total). The cluster analysis presented that these resources were divided into two main groups. There were differences in the chemical components and the contents of Asarum sieboldii from the 6 wild habitats. Except for some same components, many unique components were identified in them respectively. In addition, some components could be detected only in some populations which had smaller genetic distance. Cluster analysis shows no direct correlation between genetic distance and geographic distance of Asarum sieboldii in Qin-ba region. The accumulations of some volatile components of Asarum sieboldii are possibly related to genetic diversity.
Sigma-Aldrich
Safrole, ≥97%