Merck
  • Post-deposition treatment of an arylated-carbazole conjugated polymer for solar cell fabrication.
Post-deposition treatment of an arylated-carbazole conjugated polymer for solar cell fabrication.
Advanced materials (Deerfield Beach, Fla.) (2012-06-22)
Xiaofeng Liu, Wen Wen, Guillermo C Bazan
ABSTRACT
An arylated-carbazole conjugated polymer with a deep HOMO level has been developed. Solar cells based on blends of PCX3 and PC(71) BM show efficiency of 3.9% with a V(oc) of 0.96 V. The device performance can be improved to 5.1% by using polar solvent treatment, most likely as a result of modified interfacial properties.
Product Description
Supelco
Carbazole, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Carbazole, ≥95% (GC)
Pricing and availability is not currently available.