Merck
  • Binding of the anticancer alkaloid sanguinarine with tRNA(phe): spectroscopic and calorimetric studies.
Binding of the anticancer alkaloid sanguinarine with tRNA(phe): spectroscopic and calorimetric studies.
Journal of biomolecular structure & dynamics (2012-06-19)
Maidul Hossain, Ayesha Kabir, Gopinatha Suresh Kumar
ABSTRACT
The interaction of the natural plant alkaloid and anticancer agent sanguinarine with tRNA(phe) has been investigated by spectroscopic and calorimetric techniques. Sanguinarine iminium binds to tRNA(phe) cooperatively; alkanolamine does not bind but in presence of large tRNA(phe) concentration, a conversion from alkanolamine to iminium occurs resulting in concomitant binding of the latter. The binding affinity of the iminium to tRNA(phe) obtained from isothermal titration calorimetry was of the order of 10(5) M(-1), which is close to that evaluated from spectroscopy. The binding was driven largely by negative enthalpy and a smaller but favourable positive entropy change. The binding was dependent on the [Na(+)] concentration, but had a larger non-electrostatic contribution to the Gibbs energy. A small heat capacity value and the enthalpy-entropy compensation in the energetics of the interaction characterized the binding of the iminium form to tRNA(phe). This study confirms that the tRNA(phe) binding moiety is the iminium form of sanguinarine.
Sigma-Aldrich
Sanguinarine chloride hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.