Merck
  • Synthesis and antitumor activity of formononetin nitrogen mustard derivatives.
Synthesis and antitumor activity of formononetin nitrogen mustard derivatives.
European journal of medicinal chemistry (2012-05-29)
Jie Ren, Hua-Jin Xu, Hong Cheng, Wen-Qun Xin, Xin Chen, Kun Hu
ABSTRACT
A series of formononetin nitrogen mustard derivatives were synthesized and evaluated in vitro for their cytotoxicity against five cancer cell lines (SH-SY5Y, HCT-116, DU-145, Hela and SGC-7901). The pharmacological results showed that many of the new derivatives displayed more potent cytotoxicity than alkeran. Furthermore, compounds 6d and 6n could induce cell cycle arrest at G2/M phase and cell apoptosis.
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%