Merck
  • Development of a novel label-free amperometric immunosensor for the detection of okadaic acid.
Development of a novel label-free amperometric immunosensor for the detection of okadaic acid.
Analytica chimica acta (2012-04-10)
Akhtar Hayat, Lise Barthelmebs, Audrey Sassolas, Jean-Louis Marty
ABSTRACT
Okadaic acid (OA), a lipophilic phycotoxin is mainly produced by toxigenic dinoflagellates. The need to develop high performing methods for OA analysis able to improve the traditional ones is evident. In this work, a novel experimental methodology for label-free detection of OA was developed. Protein G magnetic beads (protein-G-MBs) modified gold electrode was used to immobilize anti-OA monoclonal antibody (anti-OA-MAb). Preliminary, colorimetric tests were performed in order to validate protein-G-MBs and anti-OA-MAb reaction. Electrochemical detection was carried out by differential pulse voltammetry in ferri/ferrocyanide solution. The limit of detection value obtained (0.5 μg L(-1)) validated the developed electrochemical immunosensor as a promising tool for routine use. The matrix effect and the recovery rate were also assessed with real samples, showing a good percentage of recovery.
Sigma-Aldrich
Potassium ferricyanide(III), powder or chunks, <10 μm, 99%
Sigma-Aldrich
Potassium hexacyanoferrate(III), ReagentPlus®, ~99%
Sigma-Aldrich
Potassium hexacyanoferrate(III), 99.98% trace metals basis
Sigma-Aldrich
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Potassium hexacyanoferrate(III), ACS reagent, ≥99.0%