Merck
  • Separation of two constituents from purple sweet potato by combination of silica gel column and high-speed counter-current chromatography.
Separation of two constituents from purple sweet potato by combination of silica gel column and high-speed counter-current chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-12-27)
Kai He, Xiaoli Ye, Xuegang Li, Hongying Chen, Lujiang Yuan, Yafei Deng, Xin Chen, Xiaoduo Li
ABSTRACT
It is known that the choice of solvent system for high speed counter-current chromatography separation is of utmost importance. In this study, a simple and rapid thin layer chromatograph coupling with fluorometric (TLC-F) method has been used to determine the partition coefficient of target compounds in HSCCC solvent system. Two components, 6,7-dimethoxycoumarin and 5-hydroxymethyl-2-furfural were successfully separated from purple sweet potato extracts by successive sample injection for the first time, using n-hexane-ethyl acetate-methanol-water (1:2:1:1, v/v/v/v) as the solvent system. Additionally, statistical analysis showed that there was no significant difference in partition coefficient obtained by the TLC-F method and by HPLC, which demonstrated the usefulness of TLC-F method.
Sigma-Aldrich
6,7-Dimethoxycoumarin, 98%