Merck
  • Superparamagnetic Fe3O4 nanoparticles-carbon nitride nanotube hybrids for highly efficient peroxidase mimetic catalysts.
Superparamagnetic Fe3O4 nanoparticles-carbon nitride nanotube hybrids for highly efficient peroxidase mimetic catalysts.
Chemical communications (Cambridge, England) (2011-11-15)
Jung Woo Lee, Hyung Joon Jeon, Hyun-Joon Shin, Jeung Ku Kang
ABSTRACT
We report a facile route to synthesize size tunable Fe(3)O(4) nanoparticles (NPs)-carbon nitride nanotube (CNNT) hybrids. These hybrids showing the water-soluble property are proven to exhibit ultra high peroxidase mimetic activity compared to those of pure NPs, where a colorless peroxidase substrate 3,3,5,5-tetramethylbenzidine changes by H(2)O(2) to its blue colored oxidized state.
Product Description
Supelco
Benzidine, analytical standard
Sigma-Aldrich
Benzidine, purum p.a., ≥98.0% (N)
Sigma-Aldrich
Benzidine dihydrochloride, ≥99% (titration)
Sigma-Aldrich
Benzidine, ≥98.0% (N)