Merck
  • Activity and stability studies of verbascoside, a novel antioxidant, in dermo-cosmetic and pharmaceutical topical formulations.
Activity and stability studies of verbascoside, a novel antioxidant, in dermo-cosmetic and pharmaceutical topical formulations.
Molecules (Basel, Switzerland) (2011-08-20)
Silvia Vertuani, Erika Beghelli, Emanuela Scalambra, Gemma Malisardi, Stefano Copetti, Roberto Dal Toso, Anna Baldisserotto, Stefano Manfredini
ABSTRACT
We here report the results of our investigations carried out on verbascoside, a phenylpropanoid glycoside known for its antioxidant, anti-inflammatory and photoprotective actions. Verbascoside was obtained from Buddleia davidii meristematic cells, obtained in turn using a sustainable biotechnology platform which employs an in vitro plant cell culture technology. Verbascoside was first investigated to assess the behaviour of the active ingredient in solution or in finished preparations, in view of its potential topical use, especially in skin protection. Stability studies were performed by HPLC, and a PCL assay was adopted to determine the radical scavenging activity toward superoxide anion. The high hydrophilic character of verbascoside, suggested in a somewhat limited range of possible applications, leading us to explore its derivatization to obtain the semi-synthetic derivative VPP, an acyl derivative of verbascoside, with an improved range of applications due to its lower hydrophilic profile. Alone, VPP revealed increased antioxidant activity, both as an active ingredient and in dermocosmetic preparations. Stability studies showed a greater stability of VPP in lipophilic vehicles, whereas the parent verbascoside proved more stable in an O/W emulsions. Verbascoside was also stable in suppositories, an interesting pharmaceutical form for possible applications in treatment of inflammation of the intestinal mucosa.
Sigma-Aldrich
Verbascoside, ≥99% (HPLC)
Verbascoside, primary reference standard