Merck
  • A new type of monoterpenoid indole alkaloid precursor from Alstonia rostrata.
A new type of monoterpenoid indole alkaloid precursor from Alstonia rostrata.
Organic letters (2011-06-22)
Xiang-Hai Cai, Mei-Fen Bao, Yu Zhang, Chun-Xia Zeng, Ya-Ping Liu, Xiao-Dong Luo
ABSTRACT
Currently, all monoterpenoid indole alkaloids (MIAs) have been derived from strictosidine, which originates from the condensation of tryptophan with secologanin in a 1:1 ratio. However, our phytochemical research on Alstonia rostrata revealed a potential new precursor for these compounds. We isolated the alstrostines A and B, and it was determined that they were derived from tryptophan and secologanin in a 1:2 ratio, which supported the presence of a new type of MIA precursor.
Product Description
Supelco
Secologanin, analytical standard
Pricing and availability is not currently available.