Merck
  • Aggregation of 2-aminobenzimidazole--a combined experimental and theoretical investigation.
Aggregation of 2-aminobenzimidazole--a combined experimental and theoretical investigation.
Chemphyschem : a European journal of chemical physics and physical chemistry (2011-05-14)
Silvia E Angelova, Milena I Spassova, Vera V Deneva, Marin I Rogojerov, Liudmil M Antonov
ABSTRACT
An investigation of 2-aminobenzimidazole was carried out by calculations at HF, MP2, and DFT levels of theory and also by UV and IR spectroscopy. The quantum chemical calculations predict a full shift of the equilibrium towards the amino form, but the absorption spectra in different solvents distinctly show a two-component equilibrium system. Examination of possible equilibria in solution shows that an equilibrium between two dimeric forms of the amino tautomer of 2-aminobenzimidazole explains the spectral observations.
Sigma-Aldrich
2-Aminobenzimidazole, 97%
Pricing and availability is not currently available.