Merck
  • Release of reduced inorganic selenium species into waters by the green fresh water algae Chlorella vulgaris.
Release of reduced inorganic selenium species into waters by the green fresh water algae Chlorella vulgaris.
Environmental science & technology (2011-02-24)
Denina Bobbie Dawn Simmons, Dirk Wallschläger
ABSTRACT
The common green fresh water algae Chlorella vulgaris was exposed to starting concentrations of 10 μg/L selenium in the form of selenate, selenite, or selenocyanate (SeCN(-)) for nine days in 10% Bold's basal medium. Uptake of selenate was more pronounced than that of selenite, and there was very little uptake of selenocyanate. Upon uptake of selenate, significant quantities of selenite and selenocyanate were produced by the algae and released back into the growth medium; no selenocyanate was released after selenite uptake. Release of the reduced metabolites after selenate exposure appeared to coincide with increasing esterase activity in solution, indicating that cell death (lysis) was the primary emission pathway. This is the first observation of biotic formation of selenocyanate and its release into waters from a nonindustrial source. The potential environmental implications of this laboratory observation are discussed with respect to the fate of selenium in impacted aquatic systems, the ecotoxicology of selenium bioaccumulation, and the interpretation of environmental selenium speciation data generated, using methods incapable of positively identifying reduced inorganic selenium species, such as selenocyanate.
Sigma-Aldrich
Potassium selenocyanate, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium selenocyanate, reagent grade, 97%