Merck
Urinary and serum metabolites of di-n-pentyl phthalate in rats.
Chemosphere (2010-10-19)
Manori J Silva, Johnathan Furr, Ella Samandar, James L Preau, L Earl Gray, Larry L Needham, Antonia M Calafat
ABSTRACT
Di-n-pentyl phthalate (DPP) is used mainly as a plasticizer in nitrocellulose. At high doses, DPP acts as a potent testicular toxicant in rats. We administered a single oral dose of 500 mg kg(-1)bw of DPP to adult female Sprague-Dawley rats (N=9) and collected 24-h urine samples 1d before and 24- and 48-h after DPP was administered to tentatively identify DPP metabolites that could be used as exposure biomarkers. At necropsy, 48 h after dosing, we also collected serum. The metabolites were extracted from urine or serum, resolved with high performance liquid chromatography, and detected by mass spectrometry. Two DPP metabolites, phthalic acid (PA) and mono(3-carboxypropyl) phthalate (MCPP), were identified by using authentic standards, whereas mono-n-pentyl phthalate (MPP), mono(4-oxopentyl) phthalate (MOPP), mono(4-hydroxypentyl) phthalate (MHPP), mono(4-carboxybutyl) phthalate (MCBP), mono(2-carboxyethyl) phthalate (MCEP), and mono-n-pentenyl phthalate (MPeP) were identified based on their full scan mass spectrometric fragmentation pattern. The ω-1 oxidation product, MHPP, was the predominant urinary metabolite of DPP. The median urinary concentrations (μg mL(-1)) of the metabolites in the first 24h urine collection after DPP administration were 993 (MHPP), 168 (MCBP), 0.2 (MCEP), 222 (MPP), 47 (MOPP), 26 (PA), 16 (MPeP), and 9 (MCPP); the concentrations of metabolites in the second 24 h urine collection after DPP administration were significantly lower than in the first collection. We identified some urinary metabolic products in the serum, but at much lower levels than in urine. Because of the similarities in metabolism of phthalates between rats and humans, based on our results and the fact that MHPP can only be formed from the metabolism of DPP, MHPP would be the most adequate DPP exposure biomarker for human exposure assessment. Nonetheless, based on the urinary levels of MHPP, our preliminary data suggest that human exposure to DPP in the United States is rather limited.
Product Description
Supelco
Dipentyl phthalate, Selectophore, ≥99.0%
Pricing and availability is not currently available.