Merck
  • Caffeine and paraxanthine HPLC assay for CYP1A2 phenotype assessment using saliva and plasma.
Caffeine and paraxanthine HPLC assay for CYP1A2 phenotype assessment using saliva and plasma.
Biomedical chromatography : BMC (2010-09-21)
Vidya Perera, Annette S Gross, Andrew J McLachlan
ABSTRACT
Caffeine has been extensively used as a probe to measure CYP1A2 activity in humans with caffeine clearance or the paraxanthine (major metabolite of caffeine) to caffeine concentration ratio being regarded as the preferred metric. A simple reverse-phased C(18) HPLC assay using ethyl acetate liquid-liquid extraction was developed to quantitate caffeine and paraxanthine concentrations in saliva and plasma. The mobile phase consisted of acetonitrile-acetic acid-H(2)O (100:1:899) and analytes were quantitated with UV detection at 280 nm. The extraction recovery for paraxanthine and caffeine was approximately 70% in both saliva and plasma. The assay was linear over the concentration ranges 0.05-2.50 and 0.05-5.00 µg/mL, for paraxanthine and caffeine, respectively, in saliva. In plasma the assay was linear over the ranges 0.025-2.50 and 0.025-5.00 µg/mL for paraxanthine and caffeine, respectively. Intra- and inter-assay precision and accuracy were less than 15%. Detection limits were 0.015 µg/mL for paraxanthine and caffeine in saliva, while it was 0.005 µg/mL for paraxanthine and caffeine in plasma. Utility was established in samples collected from two healthy volunteers who abstained from caffeine for 24 h and received a single 100 mg oral dose of caffeine. The assay developed is a robust, simple and precise technique to measure caffeine and paraxanthine in saliva and plasma of healthy volunteers after a single oral dose of caffeine.
Product Description
Supelco
Paraxanthine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1,7-Dimethylxanthine, ~98%, solid