Merck
  • Tetrathiomolybdate mediated rearrangement of aziridinemethanol tosylates: a thia-aza-Payne rearrangement.
Tetrathiomolybdate mediated rearrangement of aziridinemethanol tosylates: a thia-aza-Payne rearrangement.
The Journal of organic chemistry (2010-07-30)
Devarajulu Sureshkumar, Srinivasamurthy Koutha, Venkataraman Ganesh, Srinivasan Chandrasekaran
ABSTRACT
Aziridinemethanol sulfonate esters react with tetrathiomolybdate 1 to give thiirane derivatives as the major product and cyclic disulfides as minor product under mild reaction conditions via an unprecedented thia-aza-Payne-type rearrangement. Interestingly, when the reaction of 1 was carried out with 2-aziridino-cyclohexanol derivatives it resulted in the formation of thia-bicyclo[3.1.1]heptane or dithia-bicyclo[3.2.1]octane derivatives.
Sigma-Aldrich
Ethylene sulfide, 98%