Merck
  • Visual detection of melamine in infant formula at 0.1 ppm level based on silver nanoparticles.
Visual detection of melamine in infant formula at 0.1 ppm level based on silver nanoparticles.
The Analyst (2010-02-23)
Cuiping Han, Haibing Li
ABSTRACT
The purpose of this paper is to develop a sensitive and low-cost colorimetric method for detection of melamine using simple circuitry. The stable p-nitroaniline-modified silver nanoparticles (Ag NPs) were synthesized by a zero-length covalent coupling chemistry, which showed sensitivity and selectivity towards melamine. The melamine induces the aggregation of nanoparticles due to electron donor-acceptor interaction between melamine and p-nitroaniline at the Ag NP interface, resulting in a shift in the surface plasmon band and a consequent color change of the Ag NPs from yellow to blue. The color change was monitored using UV-vis spectrophotometry. More importantly, this optical method was successfully utilized to a practical infant formula sample. As low as 0.1 ppm melamine in infant formula could be distinguished upon a color change of solution in two minutes.
Product Description
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
4-Nitroaniline, ≥99%