Merck
  • Voltage-dependent calcium and chloride currents in S17 bone marrow stromal cell line.
Voltage-dependent calcium and chloride currents in S17 bone marrow stromal cell line.
Journal of cellular physiology (2010-01-06)
Henrique B Silva, Emiliano Medei, Deivid C Rodrigues, Edson Rondinelli, Norma A S Almeida, Regina C S Goldenberg, Antonio C Campos de Carvalho, José H M Nascimento
ABSTRACT
The bone marrow stromal cell line S17 has been used to study hematopoiesis in vitro. In this study, we demonstrate the presence of calcium and chloride currents in cultured S17 cells. Calcium currents were of low amplitude or barely detectable (50-100 pA). Hence to amplify the currents, we have used barium as a charge carrier. Barium currents were identified based on their distinct voltage-dependence, and sensitivity to dihydropyridines. S17 cells also exhibited a slowly activating outward current without inactivation, most commonly seen when the sodium of the extracellular solution was replaced either by TEA (TEA/Cs saline) or NMDG (NMDG saline), or by addition of amiloride to the extracellular solution. This current was abolished either by 500 microM SITS (4,4'-diisothiocyanatostilbene-2-2'-disulfonic acid) or 500 microM DPC (diphenylamine-2-carboxylic acid) a cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel blocker, identifying it as a Cl(-) current. RT-PCR identified the presence of ENaC and CFTR transcripts. CFTR blockade reduced cell proliferation, suggesting that this channel plays a physiological role in regulation of S17 cell proliferation.
Sigma-Aldrich
N-Phenylanthranilic acid, 98%