Merck
  • Highly enantioselective synthesis of (S)-alpha-alkyl-alpha,beta-diaminopropionic acids via asymmetric phase-transfer catalytic alkylation of 2-phenyl-2-imidazoline-4-carboxylic acid tert-butyl esters.
Highly enantioselective synthesis of (S)-alpha-alkyl-alpha,beta-diaminopropionic acids via asymmetric phase-transfer catalytic alkylation of 2-phenyl-2-imidazoline-4-carboxylic acid tert-butyl esters.
Organic letters (2009-07-29)
Yohan Park, Sukhoon Kang, Young Ju Lee, Taek-Soo Kim, Byeong-Seon Jeong, Hyeung-geun Park, Sang-sup Jew
ABSTRACT
An efficient enantioselective synthetic method for (S)-alpha-alkyl-alpha,beta-diaminopropionic acid is reported. The asymmetric phase-transfer catalytic alkylation of N(1)-Boc-2-phenyl-2-imidazoline-4-carboxylic acid tert -butyl ester in the presence of chiral quaternary ammonium catalyst gave the corresponding alkylated products (93-98% ee) which could be transformed to enantioenriched alpha-alkyl-alpha,beta-diaminopropionic acids.
Sigma-Aldrich
DL-2,3-Diaminopropionic acid monohydrochloride, 98%
Pricing and availability is not currently available.