Merck
  • Organocatalytic syn-aldol reactions of dioxanones with (S)-isoserinal hydrate: synthesis of L-deoxymannojirimycin and L-deoxyidonojirimycin.
Organocatalytic syn-aldol reactions of dioxanones with (S)-isoserinal hydrate: synthesis of L-deoxymannojirimycin and L-deoxyidonojirimycin.
The Journal of organic chemistry (2009-05-02)
Nagarjuna Palyam, Marek Majewski
ABSTRACT
We report a new protocol for synthesis of L-1-deoxymannojirimycin, L-1-deoxyidonojirimycin, and the N-isopropyl derivative of the latter compound from the readily available precursors (S)-isoserinal hydrate and 2-tert-butyl-2-methyl-1,3-dioxan-5-one. The key steps include diastereoselective proline-catalyzed syn aldol transformation and a reductive amination/cyclization.
Sigma-Aldrich
DL-Isoserine, 98%