Merck
  • Efficient synthesis of spiro[chromeno[2,3-d]pyrimidine-5,3'-indoline]-tetraones by a one-pot and three-component reaction.
Efficient synthesis of spiro[chromeno[2,3-d]pyrimidine-5,3'-indoline]-tetraones by a one-pot and three-component reaction.
Journal of combinatorial chemistry (2009-02-26)
Khosrow Jadidi, Ramin Ghahremanzadeh, Ayoob Bazgir
ABSTRACT
An efficient one-pot synthesis of novel 8,9-dihydrospiro[chromeno[2,3-d]pyrimidine-5,3'-indoline]-2,2',4,6(1H,3H,7H)-tetraone derivatives by a three-component condensation reaction of barbituric acids, isatins and cyclohexane-1,3-diones in refluxing water in the presence of p-TSA for 10 h is reported. Two cyclohexane-1,3-diones, three barbituric acids, and eight substituted isatins were chosen for the library validation. Reaction of 5,5-dimethyl-cyclohexane-1,3-dione and acenaphthylene-1,2-dione with barbituric acids resulted in the formation of spiro[acenaphthylene-1,5'-chromeno[2,3-d]pyrimidine] derivatives.
Sigma-Aldrich
1,3-Cyclohexanedione, 97%