Merck
  • Memory reconsolidation mediates the strengthening of memories by additional learning.
Memory reconsolidation mediates the strengthening of memories by additional learning.
Nature neuroscience (2008-10-14)
Jonathan L C Lee
ABSTRACT
Memories are dynamic, rather than static, in nature. The reactivation of a memory through re-exposure to salient training stimuli results in its destabilization, necessitating a restabilization process known as reconsolidation, a disruption of which leads to amnesia. I found that one normal function of hippocampal memory reconsolidation in rats is to modify the strength of a contextual-fear memory as a result of further learning.
Sigma-Aldrich
clasto-Lactacystin β-lactone