Merck
  • Use of cyclohexanedimethanol as a nonflammable organic solvent for industrial scale reversed phase chromatography.
Use of cyclohexanedimethanol as a nonflammable organic solvent for industrial scale reversed phase chromatography.
Journal of chromatography. A (2008-07-16)
Alan K Hunter, Sydney D Hoeltzli, Gary V Johnson, Mark E Gustafson, Sa V Ho
ABSTRACT
This work demonstrates that cyclohexanedimethanol is an effective nonflammable organic solvent for the pilot scale reversed phase chromatography of recombinant apolipoprotein A-I(Milano). Cyclohexanedimethanol has low viscosity in water, enabling the use of conventional low pressure process scale chromatography columns and hardware. Results from pilot scale manufacturing using a 30 cm diameter CG71M packed column indicate that a 5.7 log reduction in E. coli host cell protein with >80% yield can be obtained for the purification of apolipoprotein A-I(Milano).
Sigma-Aldrich
1,4-Cyclohexanedimethanol, mixture of cis and trans, 99%