Merck
  • A novel polyurethane-based root canal-obturation material and urethane-acrylate-based root canal sealer-part 2: evaluation of push-out bond strengths.
A novel polyurethane-based root canal-obturation material and urethane-acrylate-based root canal sealer-part 2: evaluation of push-out bond strengths.
Journal of endodontics (2008-04-26)
Bor-Shiunn Lee, Eddie Hsiang-Hua Lai, Ken-Hsuan Liao, Chung-Yi Lee, Kuo-Huang Hsieh, Chun-Pin Lin
ABSTRACT
We have developed a visible-light curable urethane-acrylate/tripropylene glycol diacrylate (UA/TPGDA) oligomer to serve as a root canal sealer and a zinc oxide/thermoplastic polyurethane (ZnO/TPU) composite to serve as a root canal obturation material. The purpose of this study was to compare the push-out bond strengths of the following 8 groups of materials: (1) Tubliseal + gutta-percha (TB/GP); (2) Tubliseal + Resilon (TB/R); (3) Epiphany + gutta-percha (EP/GP); (4) Epiphany + Resilon (EP/R); (5) EndoREZ sealer + EndoREZ cone (ES/EC); (6) EndoREZ sealer + ZnO/TPU (ES/PU); (7) UA/TPGDA + EndoREZ cone (UA/EC); and (8) UA/TPGDA + ZnO/TPU (UA/PU). Eighty 1-mm-thick root slices prepared from extracted human permanent molars were randomly divided into 8 groups with 10 specimens in each group. Root slices were filled with the above obturation materials, and then push-out test was performed with a universal testing machine. The results showed that the UA/EC and UA/PU groups had significantly higher bond strengths than the other groups.
Sigma-Aldrich
Tri(propylene glycol) diacrylate, mixture of isomers, contains MEHQ and HQ as inhibitors, technical grade
Pricing and availability is not currently available.