Merck
  • Gas-phase tautomeric equilibrium of 4-hydroxypyrimidine with its ketonic forms: a free jet millimeterwave spectroscopy study.
Gas-phase tautomeric equilibrium of 4-hydroxypyrimidine with its ketonic forms: a free jet millimeterwave spectroscopy study.
Journal of the American Chemical Society (2007-04-24)
Raquel Sanchez, B Michela Giuliano, Sonia Melandri, Laura B Favero, Walther Caminati
ABSTRACT
4-Hydroxypyrimidine (4HP) has two conformational forms (the hydroxyl hydrogen cis or trans with respect to the adjacent nitrogen), which are in tautomeric equilibrium with two ketonic forms, 4-pyrimidinone (4PO) and 6-pyrimidinone (6PO). We have investigated the free jet absorption millimeterwave spectrum of this system, assigning the rotational spectra of 4HPcis and 4PO; the latter species is more stable by 2.0(9) kJ/mol. No lines corresponding to the trans isomer of 4-hydroxypyridine and to 6PO have been observed.
Sigma-Aldrich
4(3H)-Pyrimidinone, ≥98%
Pricing and availability is not currently available.