Merck
  • The first catalytic Sakurai reaction of N-alkoxycarbonylamino sulfones with allyltrimethylsilane.
The first catalytic Sakurai reaction of N-alkoxycarbonylamino sulfones with allyltrimethylsilane.
Organic & biomolecular chemistry (2007-02-03)
Thierry Ollevier, Zhiya Li
ABSTRACT
We report the first catalytic Sakurai reaction of N-alkoxycarbonylamino sulfones with allylsilanes. The allylation reaction of N-alkoxycarbonylamino phenylsulfones with allyltrimethylsilane proceeded smoothly with low catalyst loading of bismuth triflate (2.0 mol%) to afford the corresponding protected homoallylic amines in moderate to very good yields (up to 96%).
Sigma-Aldrich
Allyltrimethylsilane, 98%
Pricing and availability is not currently available.