Merck
  • Subclinical and clinical mastitis in heifers following the use of a teat sealant precalving.
Subclinical and clinical mastitis in heifers following the use of a teat sealant precalving.
Journal of dairy science (2006-12-22)
K I Parker, C Compton, F M Anniss, A Weir, C Heuer, S McDougall
ABSTRACT
This study investigated the effect in heifers of infusion of a bismuth subnitrate teat-canal sealant and bacterial intramammary infection (IMI) precalving on prevalence of postcalving IMI and incidence of clinical mastitis in the first 2 wk postcalving. Glands (n = 1,020) from heifers (n = 255) in 5 seasonally calving, pasture-fed dairy herds were randomly assigned within heifer to 1 of 4 treatment groups (no treatment; mammary gland secretion collection; infusion of a teat sealant; or sample collection with infusion of teat sealant). Heifers within a herd were enrolled on one calendar day, 31 d on average before the planned start of the seasonal calving period. Duplicate milk samples were collected from each gland within 4 d after calving for bacterial culture. Herd owners collected duplicate milk samples, before treatment, for bacterial culture from glands they defined as having clinical mastitis. The gland prevalence of IMI precalving was 15.5% and did not differ between herds. Bacteria isolated precalving included coagulase-negative staphylococci (76.9% of all bacteriologically positive samples), Streptococcus uberis (14.1%), Staphylococcus aureus (5.1%), Corynebacterium spp. (3.8%), and others (0.1%). The presence of an IMI precalving increased the risk of an IMI postcalving 3.6-fold and the risk of clinical mastitis 4-fold, relative to no IMI precalving. Infusion of the teat sealant reduced the risk of postcalving IMI due to Strep. uberis by 84%, and of clinical mastitis by 68%. Sampling the glands precalving had no effect on postcalving IMI or on clinical mastitis incidence. Use of an internal teat canal sealant in heifers precalving may be a useful tool for reducing the risk of subclinical and clinical mastitis in heifers.
Sigma-Aldrich
Bismuth subnitrate, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth(III) subnitrate, 98%
Pricing and availability is not currently available.