Merck
  • Kinetic resolution of (+/-)-1-phenylethanol in [Bmim][PF6] using high activity preparations of lipases.
Kinetic resolution of (+/-)-1-phenylethanol in [Bmim][PF6] using high activity preparations of lipases.
Bioorganic & medicinal chemistry letters (2006-12-13)
Shweta Shah, Munishwar N Gupta
ABSTRACT
Lipases from two different sources Candida rugosa (CRL) and Burkholderia cepacia (BCL) were formulated as enzyme precipitated and rinsed with organic solvents, organic solvent rinsed enzyme preparation, cross-linked enzyme aggregates (CLEAs) and protein coated micro-crystals (PCMCs). These various enzyme formulates were evaluated for the kinetic resolution of (+/-)-1-phenylethanol in ionic liquid [Bmim][PF(6)] by transesterification with vinyl acetate. Of all the enzyme forms evaluated EPRP and PCMC in the case of CRL showed the best results with 26 % (E value=153) and 53% (E value=79) conversion, respectively, at 35 degrees C in 24h. Carrying out this conversion with PCMC at lower temperature of 25 degrees C further improved the E value to 453 (with 44% conversion in 12h). For BCL the acetone-rinsed enzyme preparation (AREP), CLEA and PCMC performed equally well with % conversion of 50 and 99 ee(p) (%) (E value >1000) in just 2h, whereas, the free lipase gave only 8% conversion.
Sigma-Aldrich
1-Phenylethanol, 98%
Sigma-Aldrich
α-Methylbenzyl alcohol, ≥99%, FCC, FG