Merck
  • Polymer-surfactant layered heterostructures by electropolymerization of phenosafranine in Langmuir-Blodgett films.
Polymer-surfactant layered heterostructures by electropolymerization of phenosafranine in Langmuir-Blodgett films.
The journal of physical chemistry. B (2006-12-01)
Shilpa N Sawant, Mukesh Doble, J V Yakhmi, S K Kulshreshtha, Akira Miyazaki, Toshiaki Enoki
ABSTRACT
Langmuir-Blodgett (LB) films of the water-soluble dye phenosafranine (PS) have been prepared by its adsorption from aqueous dye solution to an arachidic acid (AA) monolayer at the air-water interface. Atomic force microscopy (AFM) images of the LB films revealed the effect of change in pH of deposition on the degree of complexation of AA with the PS dye. Well-defined circular islands and holes were observed which disappeared with the increase in pH. Polarized absorption studies indicated that the dye molecules are oriented uniaxially with their long axis titled at a constant angle to the surface normal of the LB film. Within the restricted geometry of the LB film, the PS dye was electropolymerized to form a two-dimensional film of poly(phenosafranine) sandwiched between arachidic acid layers. The film was characterized by IR spectroscopy, cyclic voltammetry, and AFM. X-ray diffraction studies reveal the presence of a layer structure in the AA-PS LB film before and after polymerization. The polymer film showed highly anisotropic electrical conductivity of ca. 10 orders of magnitude. This indicates the formation of two-dimensional polyPS layers between arachidic acid layers resulting in a layered heterostructure film having alternate conducting and insulating regions. Also, the conductivity of the polyPS prepared from LB film was found to be approximately 2.5 times higher than the conductivity of polyPS prepared by solution polymerization method.
Sigma-Aldrich
Phenosafranin, Dye content 80 %