Merck
  • Elucidation of the redox behavior of 2,5-dimercapto-1,3,4-thiadiazole (DMcT) at poly(3,4-ethylenedioxythiophene) (PEDOT)-modified electrodes and application of the DMcT-PEDOT composite cathodes to lithium/lithium ion batteries.
Elucidation of the redox behavior of 2,5-dimercapto-1,3,4-thiadiazole (DMcT) at poly(3,4-ethylenedioxythiophene) (PEDOT)-modified electrodes and application of the DMcT-PEDOT composite cathodes to lithium/lithium ion batteries.
Langmuir : the ACS journal of surfaces and colloids (2006-11-30)
Yasuyuki Kiya, Geoffrey R Hutchison, Jay C Henderson, Tomoo Sarukawa, Osamu Hatozaki, Noboru Oyama, Héctor D Abruña
ABSTRACT
The redox reactions of DMcT at PEDOT-modified glassy carbon electrodes (GCEs) in acetonitrile (AN) have been investigated via cyclic voltammetry (CV) and the electrochemical quartz crystal microbalance (EQCM) in order to elucidate the redox reaction mechanism. A redox couple at -0.29 V versus Ag/Ag+ was assigned to the dimerization process of singly protonated DMcT (DMcT-1H), and a second couple observed at +0.42 V was assigned to the polymerization process of the protonated DMcT dimer. Our investigations revealed further that the anodic current response at +0.55 V (polymerization process) has a shoulder at +0.38 V ascribed to the dimerization process of doubly protonated DMcT (DMcT-2H), indicating that the redox couple at +0.42 V is the overlapping response of the polymerization of the protonated DMcT dimer and the dimerization of the DMcT-2H monomer. It was also confirmed that the dimerization process of DMcT-1H at -0.29 V proceeded not only at the surface of a PEDOT film but also inside the film as previously suggested. Moreover, the thermodynamics of these redox reactions at PEDOT-modified GCEs are dependent on the basicity (or acidity) of the solution, as anticipated and previously shown at unmodified GCEs. The oxidation of DMcT occurs at less positive potentials and the reduction occurs at more negative potentials in the presence of base. On the basis of the results obtained, the full redox reaction scheme for DMcT at a PEDOT-modified GCE is proposed.
Sigma-Aldrich
1,3,4-Thiadiazole-2,5-dithiol, 98%
Sigma-Aldrich
1,3,4-Thiadiazole-2,5-dithiol dipotassium salt, 98%