Merck
  • Facile aerobic photo-oxidation of aldehydes in the presence of catalytic lithium bromide.
Facile aerobic photo-oxidation of aldehydes in the presence of catalytic lithium bromide.
Chemical & pharmaceutical bulletin (2006-10-04)
Shin-ichi Hirashima, Akichika Itoh
ABSTRACT
Aldehydes were found to be oxidized to the corresponding carboxylic acid in the presence of catalytic lithium bromide under photo-irradiation.
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Pricing and availability is not currently available.