Merck
Molecular cloning of streptomyces genes encoding vanillate demethylase.
Bioscience, biotechnology, and biochemistry (2006-09-09)
Motohiro Nishimura, Daisuke Ishiyama, Julian Davies
ABSTRACT
The vanillate demethylase genes from Streptomyces sp. NL15-2K were cloned and sequenced. The vanA and vanB gene homologs, which encode the terminal oxygenase subunit (VanA) and the ferredoxin-type reductase subunit (VanB) of the enzyme respectively, were found in the sequenced 7.5-kb DNA region. Expression of the vanAB genes in Streptomyces lividans 1326 resulted in in vivo demethylation of veratric acid to vanillic acid.
Sigma-Aldrich
3,4-Dimethoxybenzoic acid, ≥99%
Pricing and availability is not currently available.