Merck
  • Effect of Ag and Au nanoparticles on the SERS of 4-aminobenzenethiol assembled on powdered copper.
Effect of Ag and Au nanoparticles on the SERS of 4-aminobenzenethiol assembled on powdered copper.
The journal of physical chemistry. B (2006-07-21)
Kwan Kim, Hyun Sook Lee
ABSTRACT
Raman scattering measurements were conducted for 4-aminobenzenethiol (4-ABT) assembled on powdered copper substrates. Initially, very weak Raman peaks were detected, but upon attaching Ag nanoparticles probably via NH2 groups onto 4-ABT/Cu, distinct Raman spectra were observed. Considering the fact that no Raman peak was identified when Ag nanoparticles were adsorbed on 4-aminophenyl-derivatized silane monolayers assembled on silica powders, the Raman spectra observed for Ag@4-ABT/Cu should be surface-enhanced Raman scattering (SERS) spectra, occurring by an electromagnetic coupling of the localized surface plasmon of Ag nanoparticles with the surface plasmon polariton of Cu powders. The extra enhancement factor attainable by the attachment of a single Ag nanoparticle is estimated to be as large as 1.4 x 10(5) in the case when 632.8-nm radiation is used as the excitation source. When Au nanoparticles were attached onto 4-ABT/Cu, at least an order of magnitude weaker Raman spectra were obtained at all excitation wavelengths, however, indicating that the Au-to-Cu coupling should be far less effective than the Ag-to-Cu coupling for the induction of SERS.
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Millipore
Nitrate Reagent B, suitable for microbiology