Merck
  • Synthesis of a nonavalent mannoside glycodendrimer based on pentaerythritol.
Synthesis of a nonavalent mannoside glycodendrimer based on pentaerythritol.
The Journal of organic chemistry (2006-02-14)
Hussein Al-Mughaid, T Bruce Grindley
ABSTRACT
A nonavalent glycodendrimer bearing terminal alpha-d-mannopyranoside units has been synthesized with a convergent approach. Terminal trivalent mannoside dendrons bearing p-halophenyl ethers were prepared by glycosylation of pentaerythritol derivatives having three 2-hydroxyethyl ether substituents. Two efficient routes were developed for the synthesis of the pentaerythritol-based core (17), which has three terminal propargyl ethers. Conditions were found under which the triple Sonogashira coupling reaction of the dendron and the tri-O-propargyl ether (17) proceeded efficiently. The product was deprotected and it and precursors were fully characterized by NMR spectroscopy and FT-ICR mass spectrometry.
Sigma-Aldrich
Pentaerythritol, 98%
Sigma-Aldrich
Pentaerythritol, 99%