Merck
  • Screening for the coccidiostats halofuginone and nicarbazin in egg and chicken muscle: development of an ELISA.
Screening for the coccidiostats halofuginone and nicarbazin in egg and chicken muscle: development of an ELISA.
Food additives and contaminants (2005-04-13)
A-C Huet, L Mortier, E Daeseleire, T Fodey, C Elliott, P Delahaut
ABSTRACT
Nicarbazin and halofuginone have been widely used as coccidiostats for the prevention and treatment of coccidiosis in poultry. It has been shown that accidental cross-contamination of feed can lead to residues of these compounds in eggs and/or muscle. This paper describes a direct competitive assay for detecting halofuginone and nicarbazin, developed as qualitative screening assay. In an optimized competitive ELISA, antibodies showed 50% binding inhibition at approximately 0.08 ng ml(-1) for halofuginone and 2.5 ng ml(-1) for dinitrocarbanilide (marker residue for nicarbazin). Extraction from the matrix was carried out with acetonitrile followed by a wash with hexane. The assay's detection capability (CCbeta) for halofuginone was < 0.5 microg kg(-1) in egg and < 1 microg kg(-1) in muscle. For dinitrocarbanilide, the CCbeta was estimated at < 3 microg kg(-1) in egg and < 10 microg kg(-1) in chicken muscle.
Product Description
Supelco
Nicarbazin, VETRANAL®, analytical standard
Sigma-Aldrich
Nicarbazin