Merck
  • Efficient plant regeneration protocol through callus for Saussurea obvallata (DC.) Edgew. (Asteraceae): effect of explant type, age and plant growth regulators.
Efficient plant regeneration protocol through callus for Saussurea obvallata (DC.) Edgew. (Asteraceae): effect of explant type, age and plant growth regulators.
Plant cell reports (2005-03-12)
Uppeandra Dhar, Mitali Joshi
ABSTRACT
A callus induction and in vitro plantlet regeneration system for the endangered state flower of Uttaranchal (Saussurea obvallata) was optimized by studying the influence of explant type (root, hypocotyl, cotyledon and leaf), age and different concentrations of plant growth regulators. Explants from 10 to 15-day-old seedlings showed maximum callus induction. Callus formation and shoot differentiation was initiated on Murashige-Skoog (MS) medium containing 6-benzyladenine (BA) and alpha-naphthalene acetic acid (NAA) in all explant types. The best results were obtained using leaf explants: 100% callusing was achieved in MS medium supplemented with 2.5 microM BA and 1.0 microM NAA, and 100% differentiation along with a multiplication rate of 12 shoots per explant with a combination of 5.0 microM BA and 1.0 microM NAA. However, the results reflected the existence of high inter-explant variability in response to growth regulators. In vitro rooting of shoots was achieved at an efficiency of 100% in one-half strength MS medium supplemented with 2.5 microM indole-3-butyric acid. Application of this protocol has potential for mass multiplication of the target species in a limited time period.
Sigma-Aldrich
1-Naphthyl acetate, ≥98% (C)