Merck
  • Rapid and selective micellar electrokinetic chromatography for simultaneous determination of amikacin, kanamycin A, and tobramycin with UV detection and application in drug formulations.
Rapid and selective micellar electrokinetic chromatography for simultaneous determination of amikacin, kanamycin A, and tobramycin with UV detection and application in drug formulations.
Electrophoresis (2005-01-26)
Hsin-Hua Yeh, Shun-Jin Lin, Ju-Yun Ko, Chia-An Chou, Su-Hwei Chen
ABSTRACT
A simple and selective micellar electrokinetic chromatography (MEKC) with UV detection is described for simultaneous determination of amikacin, tobramycin, and kanamycin A, performed in Tris buffer (180 mM; pH 9.1) with 300 mM sodium pentanesulfonate (SPS) as an anionic surfactant. Under this condition, good separation with high efficiency and the required short analysis time is achieved. The linear ranges of the method for the determination of amikacin, tobramycin, and kanamycin A were 0.1-0.5 mg / mL, 0.4-2.0 mg / mL, and 0.4-2.0 mg / mL, respectively; the detection limits (signal-to-noise ratio = 3; injection, 0.5 psi 5 s) were 0.08, 0.2, and 0.2 mg / mL, respectively. The small amount of sample required and the expeditiousness of the procedure allow content uniformity to be determined in individual commercial products.
Sigma-Aldrich
Sodium pentanesulfonate, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pentanesulfonate, ≥95% (elemental analysis)
Pricing and availability is not currently available.