Merck
  • Anti-sulfonylbenzoate antibodies as a tool for the detection of nucleotide-binding proteins for functional proteomics.
Anti-sulfonylbenzoate antibodies as a tool for the detection of nucleotide-binding proteins for functional proteomics.
Journal of proteome research (2004-12-15)
Lisa L Moore, Ashley M Fulton, Marietta L Harrison, Robert L Geahlen
ABSTRACT
Proteins that bind ATP and GTP are important cellular components. We developed an immunological approach to selectively tag nucleotide-binding proteins based on the use of 5'-[4-(fluorosulfonyl)benzoyl]adenosine and 5'-[4-(fluorosulfonyl)benzoyl]guanosine affinity tags and an antibody against 4-(sulfonyl)benzoate. Detection follows affinity labeling, gel electrophoresis, and ester bond cleavage to expose the epitope. Trial analyses of labeled proteins from lymphoid cells identified multiple ATP-binding proteins, including chaperones, actin, kinases, an RNA splicing factor, a membrane ATPase, and ATP synthase.
Sigma-Aldrich
4-(Fluorosulfonyl)benzoic acid, 95%