Merck
  • Combined ab initio computational and experimental multinuclear solid-state magnetic resonance study of phenylphosphonic acid.
Combined ab initio computational and experimental multinuclear solid-state magnetic resonance study of phenylphosphonic acid.
Magnetic resonance in chemistry : MRC (2004-04-20)
Christel Gervais, Mickaël Profeta, Vincent Lafond, Christian Bonhomme, Thierry Azaïs, Hubert Mutin, Chris J Pickard, Francesco Mauri, Florence Babonneau
ABSTRACT
1H, 13C, 17O and 31P NMR parameters, including chemical shift tensors and quadrupolar parameters for 17O, were calculated for phenylphosphonic acid, C6H5PO(OH)2, under periodic boundary conditions. The results are in very good agreement with experimental data and permit the unambiguous assignment of all the sites present in the structure. In particular, the 17O NMR parameters of the P=O and P-OH environments were precisely determined, which should help in the characterization of the bonding mode of phosphonate molecules in hybrid solids. Moreover, the effect of intermolecular interactions on the NMR parameters were investigated by comparing the results of the calculations in the crystal and in an isolated molecule of phenylphosphonic acid.
Sigma-Aldrich
Phenylphosphonic acid, 98%