Merck
  • Temporal dynamics and degradation activity of an bacterial inoculum for treating waste metal-working fluid.
Temporal dynamics and degradation activity of an bacterial inoculum for treating waste metal-working fluid.
Environmental microbiology (2004-02-12)
Christopher J van der Gast, Andrew S Whiteley, Ian P Thompson
ABSTRACT
In order for established bioreactors to be effective for treating chemically mixed wastes such as metal working fluids (MWF) it is essential that they harbour microbial populations that can maintain sufficient active biomass and degrade each of the chemical constituents present. In this study we investigated the effectiveness of a bacterial consortium composed of four species (Clavibacter michiganensis, Methylobacterium mesophilicum, Rhodococcus erythropolis and Pseudomonas putida), assembled on the basis of their apparent ubiquity in waste MWF, degradation ability and tolerance to fluctuating chemistry of the waste. The temporal dynamics of the inoculum and its effects on the fate of individual chemical components of the waste were studied, by regular sampling, over 400 h. Using a complementary approach of culture with chemotaxonomic (FAME) analysis and applying group specific probes (FISH), the inoculum was found to represent a significant component of the community in bioreactors with and without presence of indigenous MWF populations. In addition, the reduction in the COD by the consortium was approximately 85% of the total pollution load, and 30-40% more effectively than any other treatment (indigenous MWF community alone or activated sludge). Furthermore, all the chemical constituents, including the biocide (a formaldehyde release agent) demonstrated > 60% reduction. Many chemical components of the MWF proved to be recalcitrant in the other treatments. The results of this study confirm that assemblage of an inoculum, based on a comprehensive knowledge of the indigenous microbial community, in the target habitat, is a highly effective way of selecting microbial populations for bioaugmentation of bioreactors.
Sigma-Aldrich
Dodecanedioic acid, 99%