Merck
  • Combining NMR and molecular modelling in a drug delivery context: investigation of the multi-mode inclusion of a new NPY-5 antagonist bromobenzenesulfonamide into beta-cyclodextrin.
Combining NMR and molecular modelling in a drug delivery context: investigation of the multi-mode inclusion of a new NPY-5 antagonist bromobenzenesulfonamide into beta-cyclodextrin.
Bioorganic & medicinal chemistry (2004-01-16)
Gloria Uccello-Barretta, Federica Balzano, Giuseppe Sicoli, Carmen Fríglola, Ignacio Aldana, Antonio Monge, Donatella Paolino, Salvatore Guccione
ABSTRACT
NMR spectroscopic and molecular modelling methods have been employed to describe the complexation of trans-N-4-[N'-(4-chlorobenzoyl)hydrazinocarbonyl]cyclohexylmethyl-4-bromobenzenesulfonamide, a new chemotype of NPY-5 antagonist, and beta-cyclodextrin, revealing the coexistence of two different kinds of 1:1 complexes where conformational changes of the guest compound with respect to the free state are also detected.