Merck
  • Mediation of beta-endorphin by isoferulic acid to lower plasma glucose in streptozotocin-induced diabetic rats.
Mediation of beta-endorphin by isoferulic acid to lower plasma glucose in streptozotocin-induced diabetic rats.
The Journal of pharmacology and experimental therapeutics (2003-09-17)
I-Min Liu, Wang-Chuan Chen, Juei-Tang Cheng
ABSTRACT
We investigated the mechanism(s) by which isoferulic acid lowers plasma glucose levels in streptozotocin-induced diabetic rats (STZ-diabetic rats). In STZ-diabetic rats, isoferulic acid dose dependently lowered plasma glucose concentrations and increased plasma beta-endorphin-like immunoreactivity (BER). Both of these effects of isoferulic acid were abolished by pretreatment of rats with tamsulosin or 2-[2,6-dimethoxyphenoxyethyl]aminomethyl-1,4-benzodioxane hydrochloride (WB 4101) at doses sufficient to block alpha1-adrenoceptors. Also, isoferulic acid enhanced BER release from isolated rat adrenal medulla in a concentration-dependent manner that could be abolished by treatment with alpha1-adrenoceptor antagonists. Moreover, bilateral adrenalectomy in STZ-diabetic rats eliminated the activities of isoferulic acid, including the plasma glucose-lowering effect and the plasma BER-elevating effect. Naloxone and naloxonazine inhibited the plasma glucose-lowering activity of isoferulic acid at doses sufficient to block opioid mu-receptors. In contrast with the effect in wild-type diabetic mice, isoferulic acid failed to lower plasma glucose levels in opioid mu-receptor knockout diabetic mice. Treatment of STZ-diabetic rats with isoferulic acid three times in 1 day resulted in an increase in the expression of the glucose transporter subtype 4 form in soleus muscle. This effect was blocked by alpha1-adrenoceptor or opioid mu-receptor antagonists. The reduction of elevated mRNA or protein level of hepatic phosphoenolpyruvate carboxykinase was also impeded in the same groups of STZ-diabetic rats. In conclusion, our results suggest that isoferulic acid may activate alpha1-adrenoceptors to enhance the secretion of beta-endorphin, which can stimulate the opioid mu-receptors to increase glucose use or/and reduce hepatic gluconeogenesis, resulting in a decrease of plasma glucose in STZ-diabetic rats.
Sigma-Aldrich
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%